Cap sealing mats, Matrix SepraSeal® and DuraSeal®
Leverancier: THERMO MATRIX TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4465 4464 4466 4490 4463BLU 4463GRE 4463YEL 4463PUR 4463GRY 4463RED 4464GRE 4464YEL 4464BLU 4464RED 4464GRY 4464PUR 4465BLU 4465GRE 4465RED 4465PUR 4465YEL 4466BLU 4466GRE 4466GRY 4466PUR 4466RED 4466YEL 4463
479-1246EA 118 EUR
479-1246 479-1245 479-1247 MATR4490 479-1267 479-1268 479-1272 479-1270 479-1269 479-1271 479-1274 479-1278 479-1273 479-1277 479-1275 479-1276 479-1279 479-1280 479-1284 479-1283 479-1285 479-1286 479-1287 479-1288 479-1289 479-1290 479-1291 MATR4463
Cap sealing mats, Matrix SepraSeal® and DuraSeal®
Afdichtingen Afdichtingen van microtiterplaten
SepraSeal® and DuraSeal® capping systems offer a pierceable, individual tube sealing solution. They are made from thermoplastic elastomer (TPE) with colorants and are DMSO resistant.
  • Low vapour transmission rates minimises evaporation and water uptake
  • Pierceable design allows samples to be accessed without removing caps
  • TPE construction ensures sealing performance will not degrade, even after multiple piercings
The SepraSeal® capping system offers the flexibility of individual tube capping plus the ability to apply 96 caps at once. Designed forMore Product Information