5-Hydroxy-DL-tryptophan

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AC12199.1 AC12199.10
MAYBAC12199.1EA 0 EUR
MAYBAC12199.1 MAYBAC12199.10
5-Hydroxy-DL-tryptophan
5-Hydroxy-DL-tryptophan
Formule: C₁₁H₁₂N₂O₃
Smeltpunt: 301 °C
Dichtheid: 1,405 g/cm³ (20 °C)
MDL Nummer: MFCD00005651
CAS nummer: 56-69-9
EINECS: 200-284-8
Merck Index: 13,04873

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR