3,5-Difluorophenylboronic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

AC35917.1
MAYBAC35917.1EA 0 EUR
MAYBAC35917.1
3,5-Difluorophenylboronic acid
3,5-Difluorophenylboronic acid
Formule: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Nummer: MFCD01318138
CAS nummer: 156545-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR