2-Amino-5-bromopyridine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BTB03304.1 BTB03304.10 BTB03304.50
MAYBBTB03304.1EA 0 EUR
MAYBBTB03304.1 MAYBBTB03304.10 MAYBBTB03304.50
2-Amino-5-bromopyridine
2-Amino-5-bromopyridine
Formule: C₅H₅BrN₂
Kookpunt: 244 °C (1013 hPa)
Smeltpunt: 136…139 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006323
CAS nummer: 1072-97-5
EINECS: 214-019-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR