1-(2-Cyanophenyl)piperidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB03852.1 BTB03852.10 BTB03852.50
MAYBBTB03852.1EA 0 EUR
MAYBBTB03852.1 MAYBBTB03852.10 MAYBBTB03852.50
1-(2-Cyanophenyl)piperidine
1-(2-Cyanophenyl)piperidine
Formule: C₁₂H₁₄N₂
MDL Nummer: MFCD00049221
CAS nummer: 72752-52-4
EINECS: 427-330-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR