2-Bromo-4,6-di-tert-butylphenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB04137.1 BTB04137.10
MAYBBTB04137.1EA 0 EUR
MAYBBTB04137.1 MAYBBTB04137.10
2-Bromo-4,6-di-tert-butylphenol
2-Bromo-4,6-di-tert-butylphenol
Formule: C₁₄H₂₁BrO
MDL Nummer: MFCD00051597
CAS nummer: 20834-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR