4-Bromo-m-toluidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB05601.1 BTB05601.10 BTB05601.50
MAYBBTB05601.1EA 0 EUR
MAYBBTB05601.1 MAYBBTB05601.10 MAYBBTB05601.50
4-Bromo-m-toluidine
4-Bromo-m-toluidine
Formule: C₇H₈BrN
MDL Nummer: MFCD00007828
CAS nummer: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR