Monomethyl succinate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

BTB06937.1 BTB06937.10
MAYBBTB06937.1EA 0 EUR
MAYBBTB06937.1 MAYBBTB06937.10
Monomethyl succinate
Monomethyl succinate
Formule: C₅H₈O₄
MW: 132.11 g/mol
MDL Nummer: MFCD00002788
CAS nummer: 3878-55-5
EINECS: 223-408-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR