4-Chlorobenzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB07036.250 BTB07036.1
CHMPBTB07036.250EA 0 EUR
CHMPBTB07036.250 MAYBBTB07036.1
4-Chlorobenzamide
4-Chlorobenzamide
Formule: C₇H₆ClNO
MW: 155.58 g/mol
Kookpunt: 314
Smeltpunt: 179
MDL Nummer: MFCD00007993
CAS nummer: 619-56-7
EINECS: 210-601-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR