3-Amino-6-methylpyridazine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB07039.1 BTB07039.10
MAYBBTB07039.1EA 0 EUR
MAYBBTB07039.1 MAYBBTB07039.10
3-Amino-6-methylpyridazine
3-Amino-6-methylpyridazine
Formule: C₅H₇N₃
MW: 109.13 g/mol
MDL Nummer: MFCD01569899
CAS nummer: 18591-82-7
EINECS: 242-430-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR