2-Bromo-3-chloro-5-(trifluoromethyl)pyridine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BTB09453DA
MAYBBTB09453DAEA 0 EUR
MAYBBTB09453DA
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Formule: C₆H₂BrClF₃N
MDL Nummer: MFCD00153072
CAS nummer: 75806-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR