5-Bromo-1,3-benzodioxole

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CC01610.1 CC01610.10
MAYBCC01610.1EA 0 EUR
MAYBCC01610.1 MAYBCC01610.10
5-Bromo-1,3-benzodioxole
5-Bromo-1,3-benzodioxole
Formule: C₇H₅BrO₂
MDL Nummer: MFCD00005821
CAS nummer: 2635-13-4
EINECS: 220-123-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR