6-Quinoxalinecarboxylic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CC11701DA CC11701EA
MAYBCC11701DAEA 0 EUR
MAYBCC11701DA MAYBCC11701EA
6-Quinoxalinecarboxylic acid
6-Quinoxalinecarboxylic acid
Formule: C₉H₆N₂O₂
MDL Nummer: MFCD01365834
CAS nummer: 6925-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR