Thiophene-3-carbaldehyde

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

CC13104.250 CC13104.1
CHMPCC13104.250EA 0 EUR
CHMPCC13104.250 MAYBCC13104.1
Thiophene-3-carbaldehyde
Thiophene-3-carbaldehyde
Formule: C₅H₄OS
MW: 112.15 g/mol
Kookpunt: 191
Smeltpunt: –30 °C
Dichtheid: 1.28 g/cm³ (24 °C)
MDL Nummer: MFCD00005466
CAS nummer: 498-62-4
EINECS: 207-865-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR