Methyl-3-amino-5-phenylthiophene-2-carboxylate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 3-amino-5-phenylthiophene-2-carboxylate

GK00498DA GK00498EA
MAYBGK00498DAEA 0 EUR
MAYBGK00498DA MAYBGK00498EA
Methyl-3-amino-5-phenylthiophene-2-carboxylate
Methyl-3-amino-5-phenylthiophene-2-carboxylate
Formule: C₁₂H₁₁NO₂S
MW: 233.2862 g/mol
MDL Nummer: MFCD00068161
CAS nummer: 100063-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR