(tert-Butylamino)triphenylphosphonium bromide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

JFD02770.250 JFD02770.1
MAYBJFD02770.250EA 0 EUR
MAYBJFD02770.250 MAYBJFD02770.1
(tert-Butylamino)triphenylphosphonium bromide
(tert-Butylamino)triphenylphosphonium bromide
Formule: C₂₂H₂₅BrNP
CAS nummer: 799-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR