2-Chloro-3-cyanopyrazine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 3-chloropyrazine-2-carbonitrile

KM04911.250 KM04911DA
MAYBKM04911.250EA 0 EUR
MAYBKM04911.250 MAYBKM04911DA
2-Chloro-3-cyanopyrazine
2-Chloro-3-cyanopyrazine
Formule: C₅H₂ClN₃
MW: 139.5400 g/mol
MDL Nummer: MFCD00219653
CAS nummer: 55557-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR