3'-Allyl-4'-hydroxyacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 3′-Allyl-4′-hydroxyacetophenone, 1-(3-Allyl-4-Hydroxy-Phenyl)Ethanone, 1-(3-Allyl-4-Hydroxyphenyl)Ethanone, 1-[4-Hydroxy-3-(2-Propen-1-Yl)Phenyl]-Ethanone, 1-(4-Hydroxy-3-Prop-2-Enyl-Phenyl)Ethanone

KM05715DA KM05715EA
MAYBKM05715DAEA 0 EUR
MAYBKM05715DA MAYBKM05715EA
3'-Allyl-4'-hydroxyacetophenone
3'-Allyl-4'-hydroxyacetophenone
Formule: C₁₁H₁₂O₂
MDL Nummer: MFCD00068015
CAS nummer: 1132-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR