4-(Bromomethyl)benzenesulphonyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

KM07335.1 KM07335.10 KM07335.50
MAYBKM07335.1EA 0 EUR
MAYBKM07335.1 MAYBKM07335.10 MAYBKM07335.50
4-(Bromomethyl)benzenesulphonyl chloride
4-(Bromomethyl)benzenesulphonyl chloride
Formule: C₇H₆BrClO₂S
Smeltpunt: 74…76 °C
MDL Nummer: MFCD00156129
CAS nummer: 66176-39-4
UN: 3261
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR