Tris(2-methylphenyl)phosphine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

MB00147.1 MB00147EA MB00147EE
MAYBMB00147.1EA 0 EUR
MAYBMB00147.1 MAYBMB00147EA MAYBMB00147EE
Tris(2-methylphenyl)phosphine
Tris(2-methylphenyl)phosphine
Formule: C₂₁H₂₁P
Kookpunt: 187…192 °C (0,05 mmHg)
Smeltpunt: 125,5 °C
MDL Nummer: MFCD00008514
CAS nummer: 6163-58-2
EINECS: 228-193-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR