Nitrofen

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

RDP00053DA RDP00053EA RDP00053.50
MAYBRDP00053DAEA 0 EUR
MAYBRDP00053DA MAYBRDP00053EA MAYBRDP00053.50
Nitrofen
Nitrofen
Formule: C₁₂H₇Cl₂NO₃
MDL Nummer: MFCD00128026
CAS nummer: 1836-75-5
EINECS: 217-406-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR