3,4-Diethoxy-3-cyclobutene-1,2-dione

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diethyl squarate, 3,4-Diethoxycyclobut-3-ene-1,2-dione

RDP00117.1 RDP00117EA
MAYBRDP00117.1EA 0 EUR
MAYBRDP00117.1 MAYBRDP00117EA
3,4-Diethoxy-3-cyclobutene-1,2-dione
3,4-Diethoxy-3-cyclobutene-1,2-dione
Formule: C₈H₁₀O₄
MW: 170.16 g/mol
Kookpunt: 88…92 °C (0.4 mmHg)
Dichtheid: 1.15 g/cm³ (20 °C)
MDL Nummer: MFCD00001333
CAS nummer: 5231-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR