2,2,2-Trifluoroethyl methanesulphonate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

RF05504.250 RF05504.1
CHMPRF05504.250EA 0 EUR
CHMPRF05504.250 MAYBRF05504.1
2,2,2-Trifluoroethyl methanesulphonate
2,2,2-Trifluoroethyl methanesulphonate
Formule: C₃H₅F₃O₃S
MDL Nummer: MFCD00025058
CAS nummer: 25236-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR