2',4'-Dihydroxy-3'-methylacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 1-(2,4-Dihydroxy-3-methylphenyl)ethanone, 1-(2,4-Dihydroxy-3-methylphenyl)ethan-1-one

RH00145.1 RH00145.10 RH00145.50
MAYBRH00145.1EA 0 EUR
MAYBRH00145.1 MAYBRH00145.10 MAYBRH00145.50
2',4'-Dihydroxy-3'-methylacetophenone
2',4'-Dihydroxy-3'-methylacetophenone
Formule: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Nummer: MFCD00010817
CAS nummer: 10139-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR