2-Chloro-3-nitropyridine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

SB00655.1 SB00655.10 SB00655.50
MAYBSB00655.1EA 0 EUR
MAYBSB00655.1 MAYBSB00655.10 MAYBSB00655.50
2-Chloro-3-nitropyridine
2-Chloro-3-nitropyridine
Formule: C₅H₃ClN₂O₂
Kookpunt: 260 °C (1013 hPa)
Smeltpunt: 99…101 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006232
CAS nummer: 5470-18-8
EINECS: 226-799-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR