2-Bromo-5-chlorotoluene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

SB01149.250 SB01149.1
CHMPSB01149.250EA 0 EUR
CHMPSB01149.250 MAYBSB01149.1
2-Bromo-5-chlorotoluene
2-Bromo-5-chlorotoluene
Formule: C₇H₆BrCl
Kookpunt: 98 °C (1013 hPa)
Dichtheid: 1,55 g/cm³ (20 °C)
MDL Nummer: MFCD00000602
CAS nummer: 14495-51-3
EINECS: 238-503-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR