4-Chlorobenzyl alcohol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (4-Chlorophenyl)methanol

SB01197.1 SB01197EA SB01197EE
MAYBSB01197.1EA 0 EUR
MAYBSB01197.1 MAYBSB01197EA MAYBSB01197EE
4-Chlorobenzyl alcohol
4-Chlorobenzyl alcohol
Formule: C₇H₇ClO
MW: 142.58 g/mol
Kookpunt: 234
Smeltpunt: 67…70 °C
MDL Nummer: MFCD00004652
CAS nummer: 873-76-7
EINECS: 212-852-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR