Diethyl (2-cyanoethyl)malonate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diethyl 2-(2-cyanoethyl)malonate

SB01209.1 SB01209.10 SB01209.50
MAYBSB01209.1EA 0 EUR
MAYBSB01209.1 MAYBSB01209.10 MAYBSB01209.50
Diethyl (2-cyanoethyl)malonate
Diethyl (2-cyanoethyl)malonate
Formule: C₁₀H₁₅NO₄
MW: 213.23 g/mol
MDL Nummer: MFCD00001966
CAS nummer: 17216-62-5
EINECS: 241-260-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR