3,5-Difluorobenzyl alcohol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

SB01684.1 SB01684EA SB01684.50
MAYBSB01684.1EA 0 EUR
MAYBSB01684.1 MAYBSB01684EA MAYBSB01684.50
3,5-Difluorobenzyl alcohol
3,5-Difluorobenzyl alcohol
Formule: C₇H₆F₂O
Kookpunt: 97…99 °C (9 mmHg)
Dichtheid: 1,27 g/cm³ (20 °C)
MDL Nummer: MFCD00010313
CAS nummer: 79538-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR