α-Bromo-3,4-dichlorotoluene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

SB01686DA SB01686EA SB01686.50
MAYBSB01686DAEA 0 EUR
MAYBSB01686DA MAYBSB01686EA MAYBSB01686.50
α-Bromo-3,4-dichlorotoluene
α-Bromo-3,4-dichlorotoluene
Formule: C₇H₅BrCl₂
Kookpunt: 84…86 °C (0,3 mmHg)
MDL Nummer: MFCD00041363
CAS nummer: 18880-04-1
UN: 3265
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR