2-Bromo-5-fluorophenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SB01713DA SB01713EA SB01713.50
MAYBSB01713DAEA 0 EUR
MAYBSB01713DA MAYBSB01713EA MAYBSB01713.50
2-Bromo-5-fluorophenyl isothiocyanate
2-Bromo-5-fluorophenyl isothiocyanate
Formule: C₇H₃BrFNS
MDL Nummer: MFCD00276607
CAS nummer: 1027513-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR