Pyridinium chlorochromate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SB01867.1 SB01867.50
MAYBSB01867.1EA 0 EUR
MAYBSB01867.1 MAYBSB01867.50
Pyridinium chlorochromate
Pyridinium chlorochromate
Formule: C₅H₆ClCrNO₃
Smeltpunt: 205 °C
MDL Nummer: MFCD00013106
CAS nummer: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08064

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR