2-Amino-3-bromo-5-nitrobenzonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SB01878.1 SB01878.10 SB01878EE
MAYBSB01878.1EA 0 EUR
MAYBSB01878.1 MAYBSB01878.10 MAYBSB01878EE
2-Amino-3-bromo-5-nitrobenzonitrile
2-Amino-3-bromo-5-nitrobenzonitrile
Formule: C₇H₄BrN₃O₂
Smeltpunt: 233…235 °C
Dichtheid: 1,949 g/cm³ (–123 °C)
MDL Nummer: MFCD00054185
CAS nummer: 17601-94-4
EINECS: 241-574-4
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR