o-Tolyl isocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SB01904.1 SB01904.10 SB01904.50
MAYBSB01904.1EA 0 EUR
MAYBSB01904.1 MAYBSB01904.10 MAYBSB01904.50
o-Tolyl isocyanate
o-Tolyl isocyanate
Formule: C₈H₇NO
Kookpunt: 185 °C (1013 hPa)
Dichtheid: 1,07 g/cm³ (20 °C)
MDL Nummer: MFCD00002009
CAS nummer: 614-68-6
EINECS: 210-389-0
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR