ω-Bromo-4-cyanoacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SPB02347.1 SPB02347.10
MAYBSPB02347.1EA 0 EUR
MAYBSPB02347.1 MAYBSPB02347.10
ω-Bromo-4-cyanoacetophenone
ω-Bromo-4-cyanoacetophenone
Formule: C₉H₆BrNO
Kookpunt: 314 °C (1013 hPa)
Smeltpunt: 92…94 °C
MDL Nummer: MFCD00052931
CAS nummer: 20099-89-2
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR