1-(2-Methoxyphenyl)-2-thiourea

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

SPB06629.1 SPB06629.10 SPB06629.50
MAYBSPB06629.1EA 0 EUR
MAYBSPB06629.1 MAYBSPB06629.10 MAYBSPB06629.50
1-(2-Methoxyphenyl)-2-thiourea
1-(2-Methoxyphenyl)-2-thiourea
Formule: (C₆H₄(OCH₃))NC(S)NH₂
Kookpunt: 310 °C (1013 hPa)
Smeltpunt: 156 °C
MDL Nummer: MFCD00022162
CAS nummer: 1516-37-6
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR