4-(Trifluoromethoxy)benzenesulphonyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

TL00343.250 TL00343.1
CHMPTL00343.250EA 0 EUR
CHMPTL00343.250 MAYBTL00343.1
4-(Trifluoromethoxy)benzenesulphonyl chloride
4-(Trifluoromethoxy)benzenesulphonyl chloride
Formule: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Kookpunt: 120…122 °C (15 mmHg)
Dichtheid: 1.536 g/cm³ (20 °C)
MDL Nummer: MFCD00042408
CAS nummer: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR