4-(Trifluoromethoxy)benzoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

TL00376.1 TL00376.10
MAYBTL00376.1EA 0 EUR
MAYBTL00376.1 MAYBTL00376.10
4-(Trifluoromethoxy)benzoic acid
4-(Trifluoromethoxy)benzoic acid
Formule: C₈H₅F₃O₃
Kookpunt: 272 °C (1013 hPa)
Smeltpunt: 153,4 °C
MDL Nummer: MFCD00002541
CAS nummer: 330-12-1
EINECS: 206-352-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR