3-Bromobenzenesulphonamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TL00520DA TL00520EA
MAYBTL00520DAEA 0 EUR
MAYBTL00520DA MAYBTL00520EA
3-Bromobenzenesulphonamide
3-Bromobenzenesulphonamide
Formule: C₆H₆BrNO₂S
MDL Nummer: MFCD00084903
CAS nummer: 89599-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR