3,5-Bis(trifluoromethyl)benzyl alcohol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TL00661.1 TL00661EA TL00661.50
MAYBTL00661.1EA 0 EUR
MAYBTL00661.1 MAYBTL00661EA MAYBTL00661.50
3,5-Bis(trifluoromethyl)benzyl alcohol
3,5-Bis(trifluoromethyl)benzyl alcohol
Formule: C₉H₆F₆O
Kookpunt: 100 °C (14 mmHg)
Smeltpunt: 53…54 °C
MDL Nummer: MFCD00009907
CAS nummer: 32707-89-4
EINECS: 251-168-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR