Trace elements C, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25-023-CI
MDTC25-023-CIEA 66.13 EUR
MDTC25-023-CI
Trace elements C, Corning®
Media voor microbiologie Additieven en supplementen voor microbiologie
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.
Order Now
Specificaties
Environmentally Preferable