Wissers voor transport, Transwab®

Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MW170 MW171 MW172P MW173C MW173P MW172C
720-0138EA 98.3 EUR
720-0138 720-0139 720-0140 720-2201 720-2202 MEDWMW172C
Wissers voor transport, Transwab®
Wattenstaafjes Wattenstaafjes voor transport
Each transport tube contains Amies medium preferred because the inorganic buffer ensures maintenance without overgrowth.

  • Excellent recovery of both aerobic and anaerobic bacteria
  • High release of micro-organisms onto plates or into broth
  • Safety cap includes double seal, inside and outside tube
  • Colour coded caps for ease of selection
  • Pre-labelled transport tube

Certificaten: MDD, IVD, and CLSI M40.

Verpakking: Easy peel pack.

Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR