Inoculating loops, Microloops® Nichrome 5
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MW193 MW195 MW190 MW192 MW194
300-0507EA 139 EUR
300-0507 300-0509 MEDWMW190 MEDWMW192 MEDWMW194
Inoculating loops, Microloops® Nichrome 5
Uitstrijkspatels Entogen en -naalden
Microloops® are a range of inoculating loops that have been designed and moulded with precision, to ensure you can rely on them for accurate volumetric technique every time.
  • Twisted wire - reduces vibrations
  • Stable during repeated heat/cool cycles
  • Fully closed loop for secure transfer of drop
Twisted Nichrome 5 wire loops offer all the benefits of platinum, without the costs. These are available in different volumetric sizes. Nichrome 5 is the finest for repeated heating. It is made up of 80% Nickel and 20% Chromium. It
gives outstanding performance atMore Product Information