Redox combination electrode, Pt4805-DPA
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
59904203
METN59904203EA 0 EUR
METN59904203
Redox combination electrode, Pt4805-DPA
Elektroden Redox Electrodes
Pt4805-DPA is a pre-pressurised, gel-filled, low-maintenance electrode for in-line measurements of oxidation reduction potential (ORP) in chemical applications.
  • Filled with gel-reference electrolyte which is pre-pressurised to 2,5 bar (36 psi)
  • Long operational electrode lifetime: Provided with a patented silver-ion trap to prevent diaphragm fouling in sulphide-containing media
  • High accuracy: The internal over-pressure ensures a gradual electrolyte outflow efficiently cleaning the diaphragm and preventing influx of process media into the reference