Conductivity standards
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51350094
METT51350094EA 29.5 EUR
METT51350094
Conductivity standards
Standaarden Geleidbaarheidsstandaarden
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature