Bürkle Dip Samplers with Plunging Siphon
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06041-32 06041-33
MFLX06041-32EA 101 EUR
MFLX06041-32 MFLX06041-33
Bürkle Dip Samplers with Plunging Siphon
Monsternemers Monsternemers voor vloeistoffen
Simple and safe removal of liquid samples.
  • Contamination-free sampling
  • Transparent PP plastic for better visibility
  • Single-handed valve operation makes multilayer and single-point sampling easy
  • Ultrasmooth surface prevents sample, residue, and dirt accumulation
The dip samplers with plunging siphon provide simple removal of liquid samples from multiple layers or targeted zones. The transparent polypropylene (PP) samplers offer excellent viewing of samples. The reflux valve at the top of the tube and ball valve at the lower end allow you to easily openMore Product Information