Parker Hannifin Panel-Mount Needle Valves, PFA
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
98402-01 98402-02 06373-23 98402-03
MFLX98402-01EA 245 EUR
MFLX98402-01 MFLX98402-02 MFLX06373-23 MFLX98402-03
Parker Hannifin Panel-Mount Needle Valves, PFA
Ventielen Needle Valves
Easily control flow with PFA needle valves.
  • Constructed of chemically inert PFA
When used with fluoropolymer tubing, these valves complete a chemically inert fluid pathway.