Masterflex® Transfer Tubing, Versilon™ A-60-G
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06410-01 06410-02 06410-03 06410-04 06410-05 06410-06 06410-07 06410-08 06410-09 06410-10 06410-11 06410-13 06410-14 06410-15 06410-17 06410-18 06410-12 06410-16 06410-19
MFLX06410-01EA 84.1 EUR
MFLX06410-01 MFLX06410-02 MFLX06410-03 MFLX06410-04 MFLX06410-05 MFLX06410-06 MFLX06410-07 MFLX06410-08 MFLX06410-09 MFLX06410-10 MFLX06410-11 MFLX06410-13 MFLX06410-14 MFLX06410-15 MFLX06410-17 MFLX06410-18 MFLX06410-12 MFLX06410-16 MFLX06410-19
Masterflex® Transfer Tubing, Versilon™ A-60-G
Slangen Transfer Tubing
Resists chemicals and UV-light.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They areMore Product Information