PendoTECH® Single-Use Pressure Monitoring and Transmitting Systems
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX19406-02EA 6000 EUR
MFLX19406-02 MFLX19406-03 MFLX19406-04 MFLX19406-05 MFLX19406-06 MFLX19406-07 MFLX19406-08 MFLX19406-10 MFLX19406-11 MFLX19406-40 MFLX19406-41 MFLX19406-09
PendoTECH® Single-Use Pressure Monitoring and Transmitting Systems
Fluid Sensors
Single-use solution to monitor process pressure.
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Data output capability to a PC or control system
  • User-configurable Min/Max alarm set points
The PressureMAT combines a pressure monitor, data logger, alarm, and transmitter in one simple control box. Designed for use with single-use, in-line pressure sensors. Each channel takes pressure readings once per second. Process pressure is displayed on the easy-to-readMore Product Information