PendoTECH® Single-Use Temperature Sensors
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX19406-13EA 138 EUR
MFLX19406-13 MFLX19406-42 MFLX19406-43 MFLX19406-16 MFLX19406-17 MFLX19406-12
PendoTECH® Single-Use Temperature Sensors
Fluid Sensors
Eliminates cross-contamination and extensive cleaning protocols.
  • Can be sterilized by gamma irradiation
  • High sensitivity to temperature change
  • Better than ±0.2 °C accuracy in specified range of 0 to 70 °C
  • Certificate of quality included
The PendoTECH single-use temperature sensors allow for highly accurate measuring of liquid temperature within a tube in a range of 0 to 70 °C. Their unique design allows part of the product that comes in contact with the fluid to be disposable. The sensors eliminate cross-contamination and extensive cleaning protocols.More Product Information